Merck
Rational approach to discover multipotent anti-Alzheimer drugs.
Journal of medicinal chemistry (2005-01-22)
Michela Rosini, Vincenza Andrisano, Manuela Bartolini, Maria L Bolognesi, Patrizia Hrelia, Anna Minarini, Andrea Tarozzi, Carlo Melchiorre
ABSTRACT
The coupling of two different pharmacophores, each endowed with different biological properties, afforded the hybrid compound lipocrine (7), whose biological profile was markedly improved relative to those of prototypes tacrine and lipoic acid. Lipocrine is the first compound that inhibits the catalytic activity of AChE and AChE-induced amyloid-beta aggregation and protects against reactive oxygen species. Thus, it emerged as a valuable pharmacological tool to investigate Alzheimer's disease and as a promising lead compound for new anti-Alzheimer drugs.
Product Description
Supelco
(R)-(+)-α-Lipoic acid, analytical standard
Sigma-Aldrich
(R)-(+)-1,2-Dithiolane-3-pentanoic acid, 97%