Merck
  • IL23 Promotes Antimicrobial Pathways in Human Macrophages, Which Are Reduced With the IBD-Protective IL23R R381Q Variant.
IL23 Promotes Antimicrobial Pathways in Human Macrophages, Which Are Reduced With the IBD-Protective IL23R R381Q Variant.
Cellular and molecular gastroenterology and hepatology (2020-06-01)
Rui Sun, Clara Abraham
ABSTRACT
Interleukin (IL)23 is a major contributor to inflammatory bowel disease (IBD) pathogenesis and is being pursued as a therapeutic target, both through targeting IL23 alone or in combination with IL12. Unexpected trial outcomes highlight the importance of understanding the cell types through which IL23 regulates immune responses, and how IL23 and IL12 compare in these responses. Macrophages are key players in IBD, and IL23 recently was found to promote inflammatory outcomes in human macrophages. This raises the possibility that IL23 may be required for additional essential macrophage functions, in particular microbial clearance, such that either blocking the IL23 pathway or the IL23R-R381Q IBD-protective variant may reduce macrophage-mediated microbial clearance. We analyzed protein expression, signaling, bacterial uptake, and intracellular bacterial clearance in human monocyte-derived macrophages through Western blot, flow cytometry, and gentamicin protection. Autocrine/paracrine IL23 was critical for optimal levels of pattern-recognition-receptor (PRR)-induced intracellular bacterial clearance in human macrophages. Mechanisms regulated by IL23 included induction of pyruvate dehydrogenase kinase 1-dependent bacterial uptake, and up-regulation of reactive oxygen species through nicotinamide adenine dinucleotide phosphate oxidase members, nitric oxide synthase 2, and autophagy through ATG5 and ATG16L1. Complementing these pathways in IL23R-deficient macrophages restored PRR-induced bacterial uptake and clearance. Janus kinase 2, TYK2, and STAT3 were required for IL23-induced mechanisms. IL23 and IL12 induced antimicrobial pathways to similar levels in human macrophages. Relative to IL23R-R381, transfected IL23R-Q381, or monocyte-derived macrophages from IL23R-Q381 carriers showed reduced bacterial uptake and clearance. We identify that autocrine/paracrine IL23 is required for optimal PRR-enhanced macrophage bacterial uptake and intracellular bacterial clearance, define mechanisms regulating IL23R-induced bacterial clearance, and determine how the IBD-protective IL23R-R381Q variant modulates these processes.
Sigma-Aldrich
PS48, ≥98% (HPLC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%