Merck
  • LncRNA GATA6-AS1 Inhibits the Progression of Non-Small Cell Lung Cancer via Repressing microRNA-543 to Up-Regulating RKIP.
LncRNA GATA6-AS1 Inhibits the Progression of Non-Small Cell Lung Cancer via Repressing microRNA-543 to Up-Regulating RKIP.
Cancer management and research (2020-10-17)
Zixuan Gong, Xiaoyu Chen, YueChao Zhang, Cheng Liu, Zhibing Wang, Xiaoyan Xu, Jun Zhu, Tongqing Xue
ABSTRACT
Much evidence unveils the significance of long non-coding RNAs (lncRNAs) in diverse cancers. This study was designed to clarify the function and mechanism of lncRNA GATA6 antisense RNA 1 (GATA6-AS1) in the progression of non-small cell lung cancer (NSCLC). GATA6-AS1, miR-543 and Raf kinase inhibitor protein (RKIP) mRNA expressions were detected by qRT-PCR. Chi-square test was adopted to analyze the relationship between GATA6-AS1 expression and the clinicopathological parameters of NSCLC patients. NSCLC cells H1299 and H460 cells were used as overexpression or knockdown models, respectively, and cell proliferation and metastasis were determined by CCK-8 and Transwell assays. RKIP, E-cadherin, N-cadherin, STAT3, p-STAT3 expressions in NSCLC cells were detected by Western blot. The targeting relationship between GATA6-AS1 and miR-543 was confirmed by dual-luciferase reporter assay. GATA6-AS1 was significantly lowly expressed in NSCLC tissues and cell lines, and its low expression level was significantly correlated with larger tumor size and positive lymph node metastasis. GATA6-AS1 overexpression inhibited the proliferation, migration, invasion and epithelial-mesenchymal transition of NSCLC cells, while GATA6-AS1 knockdown caused the opposite effects. Mechanistically, it was confirmed that GATA6-AS1 impeded NSCLC cell proliferation and metastasis by adsorbing miR-543 and up-regulating the expression of RKIP. As a tumor suppressor, GATA6-AS1 participates in suppressing the progression of NSCLC by modulating the miR-543/RKIP axis.