Merck
  • Claudin-3 and claudin-4 expression in serous papillary, clear-cell, and endometrioid endometrial cancer.
Claudin-3 and claudin-4 expression in serous papillary, clear-cell, and endometrioid endometrial cancer.
Gynecologic oncology (2008-03-04)
Gottfried E Konecny, Rachana Agarwal, Gary A Keeney, Boris Winterhoff, Monica Brown Jones, Andrea Mariani, Darren Riehle, Christina Neuper, Sean C Dowdy, He-Jing Wang, Patrice J Morin, Karl C Podratz
ABSTRACT
Tight junction (TJ) proteins claudin-3 and claudin-4 may be differentially expressed in uterine serous papillary carcinoma (USPC), a rare form of endometrial cancer characterized by a particularly poor prognosis. Our aim was to determine the expression pattern and prognostic relevance of claudin-3 and claudin-4 in a large cohort of endometrial cancer patients of diverse histological type and stage. Claudin-3 and claudin-4 expression was studied in a cohort of 287 patients with endometrial cancer including 137 cases of USPC or clear-cell histology using immunohistochemistry. Patients were completely surgically staged. Outcome data is available on all 287 patients. The rate of claudin-3 and claudin-4 expression was significantly higher in USPC and clear-cell endometrial cancer compared to endometrioid endometrial cancer (claudin-3: 78% and 61% versus 38%, p<.0001; claudin-4: 56% and 44% versus 9%, p<.0001). Furthermore, expression of both TJ proteins was significantly associated with poor clinical outcome (claudin-3, DFS RR 1.70, p=.0087, OS RR 1.62, p=.0247; claudin-4, DFS RR 2.66, p<0.0001, and OS RR 2.50, p<0.0001). However, both markers did not maintain prognostic independence in multivariate analyses, as their expression was tightly associated with more advanced disease stages (p<.0001 for both), and higher nuclear grade (p<.0001 for both). These clinical observations confirm the hypothesis based on preclinical evidence that increased expression of claudin-3 and claudin-4 may contribute to the aggressive phenotype of endometrial cancer of serous papillary or clear-cell histology and suggest their potential utility as diagnostic biomarkers and possible targets for therapeutic intervention.
Sigma-Aldrich
Claudin-4 (EP417) Rabbit Monoclonal Primary Antibody