Merck
  • Stanniocalcin 2 contributes to aggressiveness and is a prognostic marker for oral squamous cell carcinoma.
Stanniocalcin 2 contributes to aggressiveness and is a prognostic marker for oral squamous cell carcinoma.
Experimental cell research (2020-05-24)
Andreia Ferreira do Carmo, Mauricio Rocha Dourado, Carine Ervolino de Oliveira, Débora Campanella Bastos, Catherine Bueno Domingueti, Lívia Máris Ribeiro Paranaíba, Íris Sawazaki-Calone, Gabriel Álvares Borges, Eliete Neves Silva Guerra, Renato C Casarin, Edgard Graner, Tuula A Salo, Roseana de Almeida Freitas, Hébel Cavalcanti Galvão, Ricardo D Coletta
ABSTRACT
Stanniocalcin 2 (STC2), a glycoprotein that regulates calcium and phosphate homeostasis during mineral metabolism, appears to display multiple roles in tumorigenesis and cancer progression. This study aimed to access the prognostic value of STC2 in oral squamous cell carcinoma (OSCC) and its implications in oral tumorigenesis. STC2 expression was examined in 2 independent cohorts of OSCC tissues by immunohistochemistry. A loss-of-function strategy using shRNA targeting STC2 was employed to investigate STC2 in vitro effects on proliferation, apoptosis, migration, invasion, epithelial-mesenchymal transition (EMT) and possible activation of signaling pathways. Moreover, STC2 effects were assessed in vivo in a xenograft mouse cancer model. High expression of STC2 was significantly associated with poor disease-specific survival (HR: 2.67, 95% CI: 1.37-5.21, p = 0.001) and high rate of recurrence with a hazard ratio of 2.80 (95% CI: 1.07-5.71, p = 0.03). In vitro downregulation of STC2 expression in OSCC cells attenuated proliferation, migration and invasiveness while increased apoptotic rates. In addition, the STC2 downregulation controlled EMT phenotype of OSCC cells, with regulation on E-cadherin, vimentin, Snail1, Twist and Zeb2. The reactivation of STC2 was observed in the STC2 knockdown cells in the in vivo xenograft model, and no influence on tumor growth was observed. Modulation of STC2 expression levels did not alter consistently the phosphorylation status of CREB, ERK, JNK, p38, p70 S6K, STAT3, STAT5A/B and AKT. Our findings suggest that STC2 overexpression is an independent marker of OSCC outcome and may contribute to tumor progression via regulation of proliferation, survival and invasiveness of OSCC cells.
Product Description
Millipore
MILLIPLEX MAP Multi-Pathway Magnetic Bead 9-Plex - Cell Signaling Multiplex Assay