Merck
  • Role of plasminogen activator inhibitor-1 in methotrexate-induced epithelial-mesenchymal transition in alveolar epithelial A549 cells.
Role of plasminogen activator inhibitor-1 in methotrexate-induced epithelial-mesenchymal transition in alveolar epithelial A549 cells.
Biochemical and biophysical research communications (2020-03-03)
Yohei Yamagami, Masashi Kawami, Takamichi Ojima, Sorahito Futatsugi, Ryoko Yumoto, Mikihisa Takano
ABSTRACT
There is increasing evidence that epithelial-mesenchymal transition (EMT) contributes to the development of organ fibrosis. We demonstrated that methotrexate (MTX) clearly induced EMT through the transforming growth factor (TGF)-β-related signaling pathway in human alveolar epithelial cell line, A549. However, critical factors associated with MTX-induced EMT have not yet been identified. In our study, we attempted to identify factors playing a crucial role in MTX-induced EMT in A549 cells. We focused on plasminogen activator inhibitor-1 (PAI-1) as the possible target for the prevention of MTX-induced EMT-related lung injury. Comprehensive gene expression analysis by microarray revealed that mRNA expression level of PAI-1 was clearly increased by MTX treatment. In addition, using several cloned A549 cells, we found a good correlation between MTX-induced increase in mRNA expression levels of α-smooth muscle actin (SMA), a representative EMT marker, and PAI-1. Furthermore, MTX upregulated mRNA and protein expression levels of PAI-1 in A549 cells; this upregulation was canceled by co-treatment with SB431542, a TGF-β-related signaling pathway inhibitor. Notably, tiplaxtinin, a PAI-1 inhibitor, and knockdown of urokinase-type plasminogen activator receptor (uPAR) prevented MTX-induced EMT in A549 cells. These findings indicate that MTX may induce EMT via upregulation of PAI-1 expression and interaction of PAI-1 with uPAR in A549 cells.