Merck
  • Hepatitis B subviral envelope particles use the COPII machinery for intracellular transport via selective exploitation of Sec24A and Sec23B.
Hepatitis B subviral envelope particles use the COPII machinery for intracellular transport via selective exploitation of Sec24A and Sec23B.
Cellular microbiology (2020-02-06)
Lisa Zeyen, Tatjana Döring, Jens T Stieler, Reinhild Prange
ABSTRACT
Hepatitis B virus (HBV) is a leading cause of liver disease. Its success as a human pathogen is related to the immense production of subviral envelope particles (SVPs) contributing to viral persistence by interfering with immune functions. To explore cellular pathways involved in SVP formation and egress, we investigated host-pathogen interactions. Yeast-based proteomics revealed Sec24A, a component of the coat protein complex II (COPII), as an interaction partner of the HBV envelope S domain. To understand how HBV co-opts COPII as a proviral machinery, we studied roles of key Sec proteins in HBV-expressing liver cells. Silencing of Sar1, Sec23, and Sec24, which promote COPII assembly concomitant with cargo loading, strongly diminished endoplasmic reticulum (ER) envelope export and SVP secretion. By analysing Sec paralog specificities, we unexpectedly found that the HBV envelope is a selective interaction partner of Sec24A and Sec23B whose functions could not be substituted by their related isoforms. In support, we found that HBV replication upregulated Sec24A and Sec23B transcription. Furthermore, HBV encountered the Sec24A/Sec23B complex via an interaction that involved the N-terminal half of Sec24A and a di-arginine motif of its S domain, mirroring a novel ER export code. Accordingly, an interference with the COPII/HBV cross-talk might display a tool to effectively inhibit SVP release.
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)