Merck
  • Long non-coding RNA CDKN2B-AS1 contributes to atherosclerotic plaque formation by forming RNA-DNA triplex in the CDKN2B promoter.
Long non-coding RNA CDKN2B-AS1 contributes to atherosclerotic plaque formation by forming RNA-DNA triplex in the CDKN2B promoter.
EBioMedicine (2020-04-27)
Minghui Ou, Xia Li, Shibo Zhao, Shichao Cui, Jie Tu
ABSTRACT
Atherosclerosis involves a slow process of plaque formation on the walls of arteries, and comprises a leading cause of cardiovascular disease. Long non-coding RNAs (lncRNAs) have been implicated in the pathogenesis of atherosclerosis. In this study, we aim to explore the possible involvement of lncRNA 'cyclin-dependent kinase inhibitor 2B antisense noncoding RNA' (CDKN2B-AS1) and CDKN2B in the progression of atherosclerosis. Initially, we quantified the expression of CDKN2B-AS1 in atherosclerotic plaque tissues and, in THP-1 macrophage-derived, and human primary macrophage (HPM)-derived foam cells. Next, we established a mouse model of atherosclerosis using apolipoprotein E knockout (ApoE-/-) mice, where lipid uptake, lipid accumulation, and macrophage reverse cholesterol transport (mRCT) were assessed, in order to explore the contributory role of CDKN2B-AS1 to the progression of atherosclerosis. RIP and ChIP assays were used to identify interactions between CDKN2B-AS1, CCCTC-binding factor (CTCF), enhancer of zeste homologue 2 (EZH2), and CDKN2B. Triplex formation was determined by RNA-DNA pull-down and capture assay as well as EMSA experiment. CDKN2B-AS1 showed high expression levels in atherosclerosis, whereas CDKN2B showed low expression levels. CDKN2B-AS1 accelerated lipid uptake and intracellular lipid accumulation whilst attenuating mRCT in THP-1 macrophage-derived foam cells, HPM-derived foam cells, and in the mouse model. EZH2 and CTCF were found to bind to the CDKN2B promoter region. An RNA-DNA triplex formed by CDKN2B-AS1 and CDKN2B promoter was found to recruit EZH2 and CTCF in the CDKN2B promoter region and consequently inhibit CDKN2B transcription by accelerating histone methylation. The results demonstrated that CDKN2B-AS1 promotes atherosclerotic plaque formation and inhibits mRCT in atherosclerosis by regulating CDKN2B promoter, and thereby could be a potential therapeutic target for atherosclerosis.