Merck
Matrix Linear Discriminant Analysis.
Technometrics : a journal of statistics for the physical, chemical, and engineering sciences (2020-06-12)
Wei Hu, Weining Shen, Hua Zhou, Dehan Kong
ABSTRACT
We propose a novel linear discriminant analysis (LDA) approach for the classification of high-dimensional matrix-valued data that commonly arises from imaging studies. Motivated by the equivalence of the conventional LDA and the ordinary least squares, we consider an efficient nuclear norm penalized regression that encourages a low-rank structure. Theoretical properties including a nonasymptotic risk bound and a rank consistency result are established. Simulation studies and an application to electroencephalography data show the superior performance of the proposed method over the existing approaches.
Sigma-Aldrich
Pyromellitic dianhydride, 97%
Pricing and availability is not currently available.