Merck
  • Integrated analysis reveals critical glycolytic regulators in hepatocellular carcinoma.
Integrated analysis reveals critical glycolytic regulators in hepatocellular carcinoma.
Cell communication and signaling : CCS (2020-06-25)
Chenying Lu, Shiji Fang, Qiaoyou Weng, Xiuling Lv, Miaomiao Meng, Jinyu Zhu, Liyun Zheng, Yumin Hu, Yang Gao, Xulu Wu, Jianting Mao, Bufu Tang, Zhongwei Zhao, Li Huang, Jiansong Ji
ABSTRACT
Cancer cells primarily utilize aerobic glycolysis for energy production, a phenomenon known as the Warburg effect. Increased aerobic glycolysis supports cancer cell survival and rapid proliferation and predicts a poor prognosis in cancer patients. Molecular profiles from The Cancer Genome Atlas (TCGA) cohort were used to analyze the prognostic value of glycolysis gene signature in human cancers. Gain- and loss-of-function studies were performed to key drivers implicated in hepatocellular carcinoma (HCC) glycolysis. The molecular mechanisms underlying Osteopontin (OPN)-mediated glycolysis were investigated by real-time qPCR, western blotting, immunohistochemistry, luciferase reporter assay, and xenograft and diethyl-nitrosamine (DEN)-induced HCC mouse models. Increased glycolysis predicts adverse clinical outcome in many types of human cancers, especially HCC. Then, we identified a handful of differentially expressed genes related to HCC glycolysis. Gain- and loss-of-function studies showed that OPN promotes, while SPP2, LECT2, SLC10A1, CYP3A4, HSD17B13, and IYD inhibit HCC cell glycolysis as revealed by glucose utilization, lactate production, and extracellular acidification ratio. These glycolysis-related genes exhibited significant tumor-promoting or tumor suppressive effect on HCC cells and these effects were glycolysis-dependent. Mechanistically, OPN enhanced HCC glycolysis by activating the αvβ3-NF-κB signaling. Genetic or pharmacological blockade of OPN-αvβ3 axis suppressed HCC glycolysis in xenograft tumor model and hepatocarcinogenesis induced by DEN. Our findings reveal crucial determinants for controlling the Warburg metabolism in HCC cells and provide a new insight into the oncogenic roles of OPN in HCC. Video Abstract.
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline
Sigma-Aldrich
Rat OPN ELISA Kit, for cell culture supernatants, plasma, and serum samples