Merck
  • MBNL1 regulates essential alternative RNA splicing patterns in MLL-rearranged leukemia.
MBNL1 regulates essential alternative RNA splicing patterns in MLL-rearranged leukemia.
Nature communications (2020-05-14)
Svetlana S Itskovich, Arun Gurunathan, Jason Clark, Matthew Burwinkel, Mark Wunderlich, Mikaela R Berger, Aishwarya Kulkarni, Kashish Chetal, Meenakshi Venkatasubramanian, Nathan Salomonis, Ashish R Kumar, Lynn H Lee
ABSTRACT
Despite growing awareness of the biologic features underlying MLL-rearranged leukemia, targeted therapies for this leukemia have remained elusive and clinical outcomes remain dismal. MBNL1, a protein involved in alternative splicing, is consistently overexpressed in MLL-rearranged leukemias. We found that MBNL1 loss significantly impairs propagation of murine and human MLL-rearranged leukemia in vitro and in vivo. Through transcriptomic profiling of our experimental systems, we show that in leukemic cells, MBNL1 regulates alternative splicing (predominantly intron exclusion) of several genes including those essential for MLL-rearranged leukemogenesis, such as DOT1L and SETD1A. We finally show that selective leukemic cell death is achievable with a small molecule inhibitor of MBNL1. These findings provide the basis for a new therapeutic target in MLL-rearranged leukemia and act as further validation of a burgeoning paradigm in targeted therapy, namely the disruption of cancer-specific splicing programs through the targeting of selectively essential RNA binding proteins.