Merck
  • Recurrence-Associated Long Non-coding RNA LNAPPCC Facilitates Colon Cancer Progression via Forming a Positive Feedback Loop with PCDH7.
Recurrence-Associated Long Non-coding RNA LNAPPCC Facilitates Colon Cancer Progression via Forming a Positive Feedback Loop with PCDH7.
Molecular therapy. Nucleic acids (2020-04-25)
Ting Li, Zhiqiang Li, Hongxing Wan, Xifeng Tang, Han Wang, Fang Chai, Meng Zhang, Baochun Wang
ABSTRACT
Long non-coding RNAs (lncRNAs) gradually show critical regulatory roles in many malignancies. However, the lncRNAs implicated in colon cancer recurrence are largely unknown. In this study, we searched the lncRNAs associated with metastasis and recurrence of colon cancer using GEO datasets. We focused on a novel lncRNA long non-coding RNA associated with poor prognosis of colon cancer (LNAPPCC), which is highly expressed in colon cancer. Increased expression of LNAPPCC is positively associated with metastasis, recurrence, and poor survival of colon cancer patients. LNAPPCC promotes colon cancer cell proliferation, migration, and in vivo xenograft growth and liver metastasis. Mechanistic investigations revealed that LNAPPCC binds EZH2, represses the binding of EZH2 to PCDH7 promoter, downregulates histone H3K27me3 level in PCDH7 promoter, and activates PCDH7 expression. Intriguingly, we also found that PCDH7 activates ERK/c-FOS signaling, increases the binding of c-FOS to LNAPPCC promoter, and activates LNAPPCC expression. Therefore, LNAPPCC and PCDH7 form a positive regulatory loop via EZH2 and ERK/c-FOS. The positive correlations between the expression of LNAPPCC, PCDH7, phosphorylated ERK, and phosphorylated c-FOS are detected in colon cancer tissues. Furthermore, depletion of PCDH7 or the adding of ERK inhibitor abolished the oncogenic roles of LNAPPCC in colon cancer. In summary, this study identified a novel lncRNA LNAPPCC that is highly expressed in colon cancer and associated with poor prognosis of colon cancer patients. LNAPPCC exerts oncogenic roles in colon cancer via forming a positive feedback loop with PCDH7. Targeting LNAPPCC/EZH2/PCDH7/ERK/c-FOS signaling axis represents a potential therapeutic strategy for colon cancer.
Sigma-Aldrich
T7 RNA Polymerase, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Anti-EZH2 Antibody, Upstate®, from rabbit