Merck
  • Weak gel of chitin with ionic liquid, 1-allyl-3-methylimidazolium bromide.
Weak gel of chitin with ionic liquid, 1-allyl-3-methylimidazolium bromide.
International journal of biological macromolecules (2009-05-27)
Kamalesh Prasad, Masa-aki Murakami, Yoshiro Kaneko, Akihiko Takada, Yoshifumi Nakamura, Jun-ichi Kadokawa
ABSTRACT
This paper reports the formation of weak gel of chitin with an ionic liquid, 1-allyl-3-methylimidazolium bromide (IL). When a mixture of 5% (w/w) chitin with IL was heated at 100 degrees C for 48h, the clear liquid was obtained. The experimental process was observed by the CCD camera view and the SEM analysis. From a mixture of chitin with IL in the higher concentration (7%, w/w), a more viscous material, i.e., a gel-like material was obtained. The rheological evaluations showed that both 5% (w/w) and 7% (w/w) chitins with IL behaved as weak gels.
Sigma-Aldrich
1-Allyl-3-methylimidazolium bromide, ≥97.0%