Merck
  • Absence of GP130 cytokine receptor signaling causes extended Stüve-Wiedemann syndrome.
Absence of GP130 cytokine receptor signaling causes extended Stüve-Wiedemann syndrome.
The Journal of experimental medicine (2020-01-09)
Yin-Huai Chen, Giedre Grigelioniene, Phillip T Newton, Jacob Gullander, Maria Elfving, Anna Hammarsjö, Dominyka Batkovskyte, Hessa S Alsaif, Wesam I Y Kurdi, Firdous Abdulwahab, Veerabahu Shanmugasundaram, Luke Devey, Séverine Bacrot, Jana Brodszki, Celine Huber, Ben Hamel, David Gisselsson, Nikos Papadogiannakis, Katarina Jedrycha, Barbara Gürtl-Lackner, Andrei S Chagin, Gen Nishimura, Dominik Aschenbrenner, Fowzan S Alkuraya, Arian Laurence, Valérie Cormier-Daire, Holm H Uhlig
ABSTRACT
The gene IL6ST encodes GP130, the common signal transducer of the IL-6 cytokine family consisting of 10 cytokines. Previous studies have identified cytokine-selective IL6ST defects that preserve LIF signaling. We describe three unrelated families with at least five affected individuals who presented with lethal Stüve-Wiedemann-like syndrome characterized by skeletal dysplasia and neonatal lung dysfunction with additional features such as congenital thrombocytopenia, eczematoid dermatitis, renal abnormalities, and defective acute-phase response. We identified essential loss-of-function variants in IL6ST (a homozygous nonsense variant and a homozygous intronic splice variant with exon skipping). Functional tests showed absent cellular responses to GP130-dependent cytokines including IL-6, IL-11, IL-27, oncostatin M (OSM), and leukemia inhibitory factor (LIF). Genetic reconstitution of GP130 by lentiviral transduction in patient-derived cells reversed the signaling defect. This study identifies a new genetic syndrome caused by the complete lack of signaling of a whole family of GP130-dependent cytokines in humans and highlights the importance of the LIF signaling pathway in pre- and perinatal development.