Merck
  • The Generation of Indole-2,3-quinodimethanes from the Deamination of 1,2,3,4-Tetrahydropyrrolo[3,4-b]indoles.
The Generation of Indole-2,3-quinodimethanes from the Deamination of 1,2,3,4-Tetrahydropyrrolo[3,4-b]indoles.
Molecules (Basel, Switzerland) (2020-01-16)
Alison Rinderspacher, Gordon W Gribble
ABSTRACT
A novel generation of indole-2,3-quinodimethanes via the deamination of 1,2,3,4-tetrahydropyrrolo[s3,4-b]indoles is reported.
Sigma-Aldrich
Diphenylacetic acid, 99%