Merck
  • Reconstitution of the beef heart and rat liver mitochondrial K+/H+ (Na+/H+) antiporter. Quantitation of K+ transport with the novel fluorescent probe, PBFI.
Reconstitution of the beef heart and rat liver mitochondrial K+/H+ (Na+/H+) antiporter. Quantitation of K+ transport with the novel fluorescent probe, PBFI.
The Journal of biological chemistry (1990-06-25)
P Jezek, F Mahdi, K D Garlid
ABSTRACT
New indicators for fluorescent measurement of Na+ and K+ ions should prove particularly useful for studies of reconstituted carriers of these ions. We show that PBFI, a K(+)-specific probe, provides a convenient and sensitive assay for the study of K+ uptake mediated by the reconstituted mitochondrial K+/H+ (Na+/H+) antiporter. Fluorescent measurements have enabled us for the first time to establish reconstitution of the K+/H+ (Na+/H+) antiporter from beef heart as well as from rat liver mitochondria. This technique has also enabled us to establish that dicyclohexylcarbodiimide is capable of complete inhibition of K+/H+ antiport in the reconstituted system, in accord with findings in intact mitochondria. PBFI fluorescence, which measures net K+ uptake, was essential for this corroboration, since dicyclohexylcarbodiimide is not capable of complete inhibition of 42K+/K+ or 86Rb+/Rb+ exchange, presumably because it acts selectively on proton transport within the carrier.