Merck
  • LINC01149 variant modulates MICA expression that facilitates hepatitis B virus spontaneous recovery but increases hepatocellular carcinoma risk.
LINC01149 variant modulates MICA expression that facilitates hepatitis B virus spontaneous recovery but increases hepatocellular carcinoma risk.
Oncogene (2019-11-23)
Rong Zhong, Jianbo Tian, Mingpeng Fu, Simin Ma, Li Liu, Jiaoyuan Li, Na Shen, Juntao Ke, Yang Yang, Yajie Gong, Ying Zhu, Ying Wang, Jing Gong, Jiang Chang, Ping Lei, Xiang Cheng, Kun Huang, Guanxin Shen, Xiaoping Miao
ABSTRACT
Interpreting disease-causing variants, especially in noncoding regions by genome-wide association studies (GWAS), has become one of the most challenging and demanding tasks. We hypothesized that functional lncRNAs variants in GWAS-identified loci might alter expression level of genes associated with persistent HBV infection and hepatocellular carcinoma (HCC). Integrated bioinformatics approaches were used to prioritize potentially functional variants and a two-stage case-control study (2473 HBV positive HCC patients, 2248 persistent HBV carriers and 2294 spontaneously recovered subjects) was performed to assess the roles of these variants. The rs2844512 G > C variant in LINC01149 was identified to facilitate HBV spontaneous recovery (OR = 0.84, 95% CI = 0.77-0.92) but increase the risk of HCC (OR = 1.21, 95% CI = 1.11-1.32) in combined samples. Subsequent biological assays indicated this variant created a binding site for miR-128-3p and upregulated MICA expression by serving as a miRNA sponge, which might recruit NK-cells to lyse infected cells, but release highly soluble MICA by shedding to induce NK-cells exhaustion and tumor immune evasion. These findings highlight a regulatory circuit between LINC01149 and MICA, mediating by miR-128-3p, and the important role of upregulated MICA in conferring susceptibility to persistent HBV infection and HCC.
Sigma-Aldrich
Human MICA ELISA Kit, for serum, plasma, cell culture supernatant and urine
Pricing and availability is not currently available.