Merck
  • LncRNA UCA1 Suppresses the Inflammation Via Modulating miR-203-Mediated Regulation of MEF2C/NF-κB Signaling Pathway in Epilepsy.
LncRNA UCA1 Suppresses the Inflammation Via Modulating miR-203-Mediated Regulation of MEF2C/NF-κB Signaling Pathway in Epilepsy.
Neurochemical research (2020-02-15)
Qian Yu, Meng-Wen Zhao, Pu Yang
ABSTRACT
Although many advances have been made in the pathogenesis of epilepsy recently, the pathological mechanisms of epilepsy are still largely unknown. Exploring the pathological mechanisms and developing novel therapeutic strategies for epilepsy are urgently needed. A SD rat model of epilepsy was established with lithium chloride-pilocarpine. Astrocytes were isolated, cultured from 8 to 12 week rats and identified by flow cytometry and immunofluorescence. Immunohistochemical staining was used for MEF2C and NF-κB in paraffin-embedded sections. RT-qPCR and western blot were used to analyze gene expression. ELISA was used to analyze the concentration of IL-6, TNF-α and Cox-2. Cells were transfected with pcDNA-MEFC2, sh-MEFC2, pcDNA-UCA1, sh-UCA1, miR-203 mimic or miR-203 inhibitor. Cell viability was assessed by MTT assay. Dual luciferase assay was used to determine the direct interaction of lncRNA UCA1/miR-203 and miR-203/MEF2C. MEF2C was down-regulated and inhibited NF-κB expression and the secretion of IL-6 and TNF-α in epilepsy. LncRNA UCA1 was also down-regulated in epilepsy. LncRNA UCA1 over-expression increased the expression of MEF2C and its knock-down decreased MEF2C expression. Luciferase activity showed lncRNA UCA1 directly targeted miR-203 and miR-203 directly targeted MEF2C. MiR-203 suppressed the expression of MEF2C, and promoted NF-κB, phosphorylated IκB/IKK and inflammatory effectors, which was reversed by MEF2C knock-down. Moreover, lncRNA UCA1 could increase the expression of MEF2C to inhibit NF-κB, phosphorylated IκB/IKK and inflammatory effectors, which was also reversed by miR-203 mimic transfection. LncRNA UCA1 inhibited the inflammation via regulating miR-203 mediated regulation of MEF2C/NF-κB signaling in epilepsy. Our investigation elucidated novel pathological mechanisms and provided potential therapeutic targets for epilepsy.