Merck
  • Benzodiazepines Drive Alteration of Chromatin at the Integrated HIV-1 LTR.
Benzodiazepines Drive Alteration of Chromatin at the Integrated HIV-1 LTR.
Viruses (2020-02-14)
Weam Elbezanti, Angel Lin, Alexis Schirling, Alexandria Jackson, Matthew Marshall, Rachel Van Duyne, Frank Maldarelli, Luca Sardo, Zachary Klase
ABSTRACT
Antiretroviral therapy (ART) lowers human immunodeficiency virus type 1 (HIV-1) viral load to undetectable levels, but does not eliminate the latent reservoir. One of the factors controlling the latent reservoir is transcriptional silencing of the integrated HIV-1 long terminal repeat (LTR). The molecular mechanisms that control HIV-1 transcription are not completely understood. We have previously shown that RUNX1, a host transcription factor, may play a role in the establishment and maintenance of HIV-1 latency. Prior work has demonstrated that inhibition of RUNX1 by the benzodiazepine (BDZ) Ro5-3335 synergizes with suberanilohydroxamic acid (SAHA) to activate HIV-1 transcription. In this current work, we examine the effect of RUNX1 inhibition on the chromatin state of the integrated HIV-1 LTR. Using chromatin immunoprecipitation (ChIP), we found that Ro5-3335 significantly increased the occupancy of STAT5 at the HIV-1 LTR. We also screened other BDZs for their ability to regulate HIV-1 transcription and demonstrate their ability to increase transcription and alter chromatin at the LTR without negatively affecting Tat activity. These findings shed further light on the mechanism by which RUNX proteins control HIV-1 transcription and suggest that BDZ compounds might be useful in activating HIV-1 transcription through STAT5 recruitment to the HIV-1 LTR.
Product Description
Alprazolam, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
SAHA, ≥98% (HPLC)