Merck
  • Overexpression of a Zn2+-sensitive soluble exopolyphosphatase from Trypanosoma cruzi depletes polyphosphate and affects osmoregulation.
Overexpression of a Zn2+-sensitive soluble exopolyphosphatase from Trypanosoma cruzi depletes polyphosphate and affects osmoregulation.
The Journal of biological chemistry (2007-09-11)
Jianmin Fang, Felix A Ruiz, Melissa Docampo, Shuhong Luo, Juliany C F Rodrigues, Lucimar S Motta, Peter Rohloff, Roberto Docampo
ABSTRACT
We report the cloning, expression, purification, and characterization of the Trypanosoma cruzi exopolyphosphatase (TcPPX). The product of this gene (TcPPX), has 383 amino acids and a molecular mass of 43.1 kDa. TcPPX differs from most exopolyphosphatases in its preference for short-chain polyphosphate (poly P). Heterologous expression of TcPPX in Escherichia coli produced a functional enzyme that had a neutral optimum pH and was dramatically inhibited by low concentrations of Zn2+, high concentrations of basic amino acids (lysine and arginine), and heparin. TcPPX is a processive enzyme and does not hydrolyze ATP, pyrophosphate, or p-nitrophenyl phosphate, although it hydrolyzes guanosine 5'-tetraphosphate very efficiently. Overexpression of TcPPX resulted in a dramatic decrease in total short-chain poly P and partial decrease in long-chain poly P. This was accompanied by a delayed regulatory volume decrease after hyposmotic stress. These results support the role of poly P in T. cruzi osmoregulation.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution