Merck
  • Eutherian-Specific Gene TRIML2 Attenuates Inflammation in the Evolution of Placentation.
Eutherian-Specific Gene TRIML2 Attenuates Inflammation in the Evolution of Placentation.
Molecular biology and evolution (2019-10-22)
Xuzhe Zhang, Mihaela Pavlicev, Helen N Jones, Louis J Muglia
ABSTRACT
Evolution of highly invasive placentation in the stem lineage of eutherians and subsequent extension of pregnancy set eutherians apart from other mammals, that is, marsupials with short-lived placentas, and oviparous monotremes. Recent studies suggest that eutherian implantation evolved from marsupial attachment reaction, an inflammatory process induced by the direct contact of fetal placenta with maternal endometrium after the breakdown of the shell coat, and shortly before the onset of parturition. Unique to eutherians, a dramatic downregulation of inflammation after implantation prevents the onset of premature parturition, and is critical for the maintenance of gestation. This downregulation likely involved evolutionary changes on maternal as well as fetal/placental side. Tripartite-motif family-like2 (TRIML2) only exists in eutherian genomes and shows preferential expression in preimplantation embryos, and trophoblast-derived structures, such as chorion and placental disc. Comparative genomic evidence supports that TRIML2 originated from a gene duplication event in the stem lineage of Eutheria that also gave rise to eutherian TRIML1. Compared with TRIML1, TRIML2 lost the catalytic RING domain of E3 ligase. However, only TRIML2 is induced in human choriocarcinoma cell line JEG3 with poly(I:C) treatment to simulate inflammation during viral infection. Its knockdown increases the production of proinflammatory cytokines and reduces trophoblast survival during poly(I:C) stimulation, while its overexpression reduces proinflammatory cytokine production, supporting TRIML2's role as a regulatory inhibitor of the inflammatory pathways in trophoblasts. TRIML2's potential virus-interacting PRY/SPRY domain shows significant signature of selection, suggesting its contribution to the evolution of eutherian-specific inflammation regulation during placentation.
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Chorionic Gonadotropin, Human Urine, Standard Grade, Chorionic Gonadotropin, Human Urine, Standard Grade, CAS 9002-61-3, is a native glycoprotein hormone that is synthesized by chorionic tissue of the placenta and found in urine during pregnancy.