Merck
  • Synthesis and Structure-Activity Relationship Study of 1-Phenyl-1-(quinazolin-4-yl)ethanols as Anticancer Agents.
Synthesis and Structure-Activity Relationship Study of 1-Phenyl-1-(quinazolin-4-yl)ethanols as Anticancer Agents.
ACS medicinal chemistry letters (2015-03-31)
Kenta Kuroiwa, Hirosuke Ishii, Kenji Matsuno, Akira Asai, Yumiko Suzuki
ABSTRACT
A quinazoline derivative PVHD121 (1a) was shown to have strong antiproliferative activity against various tumor-derived cell lines, including A549 (lung), NCI-H460 (lung), HCT116 (colon), MCF7 (breast), PC3 (prostate), and HeLa (cervical) cells with IC50 values from 0.1 to 0.3 μM. A structure-activity relationship (SAR) study at the 2- and 4-position of the quinazoline core lead to the discovery of more potent anticancer agents (14, 16, 17, 19, 24, and 31). The results of an in vitro tubulin polymerization assay and fluorescent-based colchicine site competition assay with purified tubulin indicated that 1a inhibits tubulin polymerization by binding to the colchicine site.
Sigma-Aldrich
PVHD121, ≥98% (HPLC)