Merck
  • Effects of hydroxycinnamic acids on the reduction of furan and α-dicarbonyl compounds.
Effects of hydroxycinnamic acids on the reduction of furan and α-dicarbonyl compounds.
Food chemistry (2020-01-04)
Sang Mi Lee, Li Wei Zheng, Youngae Jung, Geum-Sook Hwang, Young-Suk Kim
ABSTRACT
The effects of hydroxycinnamic acids such as cinnamic acid (CNA), p-coumaric acid(CMA), caffeic acid (CFA), and chlorogenic acid (CGA) on the reduction of furan in canned-coffee model systems (CCMS) containing α-dicarbonyls [glyoxal (GO) or methylglyoxal (MGO)] were investigated. The concentration of furan in CCMS containing GO, which was 59.76 μg/L, was reduced by the addition of CFA and CGA to 48.31 μg/L and 41.38 μg/L, respectively; similarly, the furan concentration in model system containing MGO was 45.79 μg/L, and this decreased to 35.41 μg/L (by CFA) and 32.65 μg/L (by CGA), respectively. In addition, the effects of hydroxycinnamic acids on the trapping of GO and MGO were determined. CFA and CGA greatly reduced the concentration of GO to 303.51 μg/L and 267.80 μg/L, respectively (compared to 515.79 μg/L in the control), whereas that of MGO was decreased to 207.01 μg/L and 219.14 μg/L (compared to 417.14 μg/L in the control). The trapping of α-dicarbonyls such as GO and MGO by CFA and CGA could be closely related to furan reduction in CCMS.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Furan, ≥99%
Sigma-Aldrich
Quinoxaline, ≥98%