Merck
  • CX-5461 Inhibits Pancreatic Ductal Adenocarcinoma Cell Growth, Migration and Induces DNA Damage.
CX-5461 Inhibits Pancreatic Ductal Adenocarcinoma Cell Growth, Migration and Induces DNA Damage.
Molecules (Basel, Switzerland) (2019-12-11)
Btissame El Hassouni, Giulia Mantini, Benoît Immordino, Godefridus J Peters, Elisa Giovannetti
ABSTRACT
Inhibition of ribosome biogenesis has recently emerged as a promising strategy for the treatment of metastatic tumors. The RNA polymerase I inhibitor CX-5461 has shown efficacy in a panel of cancer types and is currently being tested in clinical trials. However, further preclinical studies to unravel molecular mechanisms underlying the activity of this drug are warranted. In this study, we have investigated the effects of CX-5461 on cell growth and migration of pancreatic cancer cells by the sulforhodamine-B and wound healing assay, respectively. Furthermore, we assessed the expression of epithelial-to-mesenchymal transition (EMT) genes by qRT-PCR, while protein expression of DNA damage marker phospho-H2A.X was studied by Western blot and immunofluorescence. CX-5461 inhibits pancreatic cancer cell growth in the nanomolar range and inhibits the migratory capability of the cells. Additionally, CX-5461 induced expression of EMT factor SNAI1 and caused DNA double-strand breaks as measured by increased expression of phospho-H2A.X. This study demonstrated that CX-5461 is active against pancreatic cancer cells and modulation of EMT factors, as well as increased expression of phospho-H2A.X, support further pre-/clinical investigations, including the analyses of these markers.
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
SAFC
Triton X-100, EMPROVE® EXPERT Ph Eur