Merck
  • Clostridioides difficile Infection Induces an Inferior IgG Response to That Induced by Immunization and Is Associated with a Lack of T Follicular Helper Cell and Memory B Cell Expansion.
Clostridioides difficile Infection Induces an Inferior IgG Response to That Induced by Immunization and Is Associated with a Lack of T Follicular Helper Cell and Memory B Cell Expansion.
Infection and immunity (2019-12-25)
Souwelimatou Amadou Amani, Tyler Shadid, Jimmy D Ballard, Mark L Lang
ABSTRACT
The intracellularly active bacterial toxin TcdB is a major Clostridioides difficile virulence factor that contributes to inflammation and tissue damage during disease. Immunization with an inactive TcdB fragment prevents C. difficile infection (CDI)-associated pathology. The protective immune response against inactive TcdB involves development of antigen-specific memory B cells and long-lived plasma cells that encode TcdB-neutralizing antibodies. Unlike the response to inactive TcdB, very little is known about the host humoral immune response to C. difficile and TcdB during primary and recurrent infection. Here, we used a murine model of C. difficile disease recurrence to demonstrate that an initial infection induced a serum IgM and mucosal IgA response against the toxin, but a low serum IgG response, which is associated with a lack of protection against disease during reinfection. Infection induced a partial expansion of the T follicular helper cell compartment, essential for B cell memory responses, and, consistent with that, failed to significantly expand the memory B cell compartment. Further, infection failed to stimulate the memory B cell compartment in preimmunized mice, although they were protected against associated disease. These results delineate the key humoral immune events that follow primary and recurrent C. difficile infection and provide a compelling inverse correlation between B cell memory and disease recurrence.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Cefoperazone sodium salt, 870 - 1015 μg/mg anhydrous basis