Merck
  • Comparative Proteome Analysis of Epicardial and Subcutaneous Adipose Tissues from Patients with or without Coronary Artery Disease.
Comparative Proteome Analysis of Epicardial and Subcutaneous Adipose Tissues from Patients with or without Coronary Artery Disease.
International journal of endocrinology (2019-09-20)
Yu Xing Zhao, Hui Juan Zhu, Hui Pan, Xue Mei Liu, Lin Jie Wang, Hong Bo Yang, Nai Shi Li, Feng Ying Gong, Wei Sun, Yong Zeng
ABSTRACT
Owing to its unique anatomical structure and metabolism, epicardial adipose tissue (EAT) has attracted amount of attention in coronary artery disease (CAD) research. Here, we analyzed differences in proteome composition in epicardial (EAT) and subcutaneous adipose tissues (SAT) from patients with or without CAD. EAT and SAT samples were collected from 6 CAD patients and 6 non-CAD patients. Isobaric Tagging for Relative and Absolute Quantitation (iTRAQ) analysis combined with liquid chromatography tandem-mass spectrometry (LC-MS/MS) was performed to identify the differentially expressed proteins. In total, 2348 proteins expressed in EAT and 2347 proteins expressed in SAT were separately identified. 385 differentially expressed proteins were found in EAT and 210 proteins were found in SAT in CAD patients compared to non-CAD patients. Many proteins differentially expressed in EAT of CAD patients were involved in biological functions associated with CAD development such as cell-to-cell signaling and interaction, inflammatory response, and lipid metabolism. Differential expressions of proteins (MMP9, S100A9, and clusterin) in EAT or SAT were involved in several signaling pathways such as mitochondrial dysfunction, acute phase inflammation, and LXR/RXR activation, which was confirmed by western blotting, and similar results were obtained. The largest profiles of differentially expressed proteins in EAT and SAT between CAD patients and non-CAD patients were identified. The significant signal pathways, mitochondrial dysfunction, and LXR/RXR activation, which differential proteins were involved in, were firstly found to play roles in EAT of CAD patients, and clusterin was firstly found to be upregulated in EAT of CAD patients.