Merck
  • TLR-stimulated IRAKM activates caspase-8 inflammasome in microglia and promotes neuroinflammation.
TLR-stimulated IRAKM activates caspase-8 inflammasome in microglia and promotes neuroinflammation.
The Journal of clinical investigation (2018-10-30)
Cun-Jin Zhang, Meiling Jiang, Hao Zhou, Weiwei Liu, Chenhui Wang, Zizhen Kang, Bing Han, Quanri Zhang, Xing Chen, Jianxin Xiao, Amanda Fisher, William J Kaiser, Masanori A Murayama, Yoichiro Iwakura, Ji Gao, Julie Carman, Ashok Dongre, George Dubyak, Derek W Abbott, Fu-Dong Shi, Richard M Ransohoff, Xiaoxia Li
ABSTRACT
NLRP3 inflammasome plays a critical spatiotemporal role in the pathogenesis of experimental autoimmune encephalomyelitis (EAE). This study reports a mechanistic insight into noncanonical NLRP3 inflammasome activation in microglia for the effector stage of EAE. Microglia-specific deficiency of ASC (apoptosis-associated speck-like protein containing a C-terminal caspase-activation and recruitment [CARD] domain) attenuated T cell expansion and neutrophil recruitment during EAE pathogenesis. Mechanistically, TLR stimulation led to IRAKM-caspase-8-ASC complex formation, resulting in the activation of caspase-8 and IL-1β release in microglia. Noncanonical inflammasome-derived IL-1β produced by microglia in the CNS helped to expand the microglia population in an autocrine manner and amplified the production of inflammatory cytokines/chemokines. Furthermore, active caspase-8 was markedly increased in the microglia in the brain tissue from patients with multiple sclerosis. Taken together, our study suggests that microglia-derived IL-1β via noncanonical caspase-8-dependent inflammasome is necessary for microglia to exert their pathogenic role during CNS inflammation.
Sigma-Aldrich
Anti-ASC Antibody, clone 2EI-7, clone 2EI-7, from mouse