Merck
  • Oct4 mediates Müller glia reprogramming and cell cycle exit during retina regeneration in zebrafish.
Oct4 mediates Müller glia reprogramming and cell cycle exit during retina regeneration in zebrafish.
Life science alliance (2019-10-09)
Poonam Sharma, Shivangi Gupta, Mansi Chaudhary, Soumitra Mitra, Bindia Chawla, Mohammad Anwar Khursheed, Rajesh Ramachandran
ABSTRACT
Octamer-binding transcription factor 4 (Oct4, also known as Pou5F3) is an essential pluripotency-inducing factor, governing a plethora of biological functions during cellular reprogramming. Retina regeneration in zebrafish involves reprogramming of Müller glia (MG) into a proliferating population of progenitors (MGPCs) with stem cell-like characteristics, along with up-regulation of pluripotency-inducing factors. However, the significance of Oct4 during retina regeneration remains elusive. In this study, we show an early panretinal induction of Oct4, which is essential for MG reprogramming through the regulation of several regeneration-associated factors such as Ascl1a, Lin28a, Sox2, Zeb, E-cadherin, and various miRNAs, namely, let-7a, miR-200a/miR-200b, and miR-143/miR-145 We also show the crucial roles played by Oct4 during cell cycle exit of MGPCs in collaboration with members of nucleosome remodeling and deacetylase complex such as Hdac1. Notably, Oct4 regulates Tgf-β signaling negatively during MG reprogramming, and positively to cause cycle exit of MGPCs. Our study reveals unique mechanistic involvement of Oct4, during MG reprogramming and cell cycle exit in zebrafish, which may also account for the inefficient retina regeneration in mammals.
Sigma-Aldrich
Anti-gapdh antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Oct-4 Antibody, Chemicon®, from rabbit