Merck
  • Potentiation of interleukin 1 alpha mediated antitumor effects by ketoconazole.
Potentiation of interleukin 1 alpha mediated antitumor effects by ketoconazole.
Cancer research (1990-08-01)
P G Braunschweiger, N Kumar, I Constantinidis, J P Wehrle, J D Glickson, C S Johnson, P Furmanski
ABSTRACT
In the present studies, the regulatory role of adrenal hormones on the antitumor activity of recombinant human interleukin 1 alpha (IL-1 alpha) was investigated. Ketoconazole, a potent but transient inhibitor of adrenal steroid hormone biosynthesis, inhibited IL-1 alpha induced increases in plasma corticosterone. In s.c. RIF-1 tumors (C3H/HeJ mice) ketoconazole potentiated IL-1 alpha induced hemorrhagic necrosis (59Fe labeled RBC uptake) and prolonged intervals of low tumor perfusion (86Rb+ uptake) and attendant depletion of tumor high energy phosphate reserves as determined by in vivo 31P nuclear magnetic resonance spectroscopy. In normal muscle and skin the ketoconazole-IL-1 alpha combination had no effect on RBC content and little or no effect on tissue perfusion. Ketoconazole potentiation of IL-1 alpha induced tumor pathophysiologies was accompanied by time and ketoconazole dose dependent potentiation of RIF-1 tumor clonogenic cell killing. Although ketoconazole at 40 mg/kg and IL-1 alpha at 25 micrograms/kg alone each produced approximately 50% clonogenic cell kill, a combined treatment (IL-1 alpha 1 h after ketoconazole) resulted in surviving fractions of approximately 1.5%. In vitro, ketoconazole and IL-1 alpha induced only additive clonogenic cell kill in primary RIF-1 explant cultures. The effect of elevated plasma corticosterone levels, induced by ketamine-acepromazine anesthesia, on IL-1 alpha responsiveness was also studied in the RIF-1 tumor model. In C3H/HeJ mice, anesthesia increased plasma corticosterone levels within 30 min, abrogated the IL-1 alpha effect on tumor perfusion, and prevented depletion of tumor high energy phosphate metabolite reserves. Our results are consistent with the hypothesis that IL-1 alpha mediated adrenal hormone responses exert a profound negative feedback on IL-1 alpha antitumor activities. Our data also indicate that adrenal steroid hormone biosynthetic pathways could provide a focus for modulation strategies to increase the efficacy of cytokine based therapeutic interventions.
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)