- Dihydroxyacetone induces G2/M arrest and apoptotic cell death in A375P melanoma cells.
Dihydroxyacetone induces G2/M arrest and apoptotic cell death in A375P melanoma cells.
The active ingredient in sunless tanning products (STPs) is a simple sugar, dihydroxyacetone (DHA). Several studies have demonstrated that DHA is absorbed within the viable layers of skin and not fully contained within the stratum corneum. Additionally, spray tanning and other aerosolized application methods have increased the risk of internal exposure through mucous membranes and inhalation. Beyond its presence in STPs, DHA also occurs as an endogenous by-product of fructose metabolism, and an excess of DHA in cells can induce advanced glycation end (AGE) products and oxidative stress. Therefore, exogenous and endogenous exposures to DHA may be harmful to cells, and it has already been demonstrated that exogenous exposure to DHA is cytotoxic in immortalized keratinocytes. Still, little is known about the exogenous DHA exposure effects on other skin components. In this study, we explore the effects of exogenous DHA exposure in a human melanoma cell line, A375P. Melanoma cells were sensitive to DHA and displayed a transient burst of reactive oxygen species within an hour of exposure. Cell cycle arrest at G2/M was observed within 24 h of exposure, and apoptosis, monitored by the cleavage of PARP-1 and Caspase-3, was detected within 72 h of exposure to DHA. Together, these demonstrate that exogenous exposure to DHA has cytotoxic effects in our selected cell model and indicates the need to further investigate the exogenous exposure effects of DHA in other relevant exposure models.