Merck
  • Constitutive interferon signaling maintains critical threshold of MLKL expression to license necroptosis.
Constitutive interferon signaling maintains critical threshold of MLKL expression to license necroptosis.
Cell death and differentiation (2018-05-23)
Joseph Sarhan, Beiyun C Liu, Hayley I Muendlein, Chi G Weindel, Irina Smirnova, Amy Y Tang, Vladimir Ilyukha, Maxim Sorokin, Anton Buzdin, Katherine A Fitzgerald, Alexander Poltorak
ABSTRACT
Interferons (IFNs) are critical determinants in immune-competence and autoimmunity, and are endogenously regulated by a low-level constitutive feedback loop. However, little is known about the functions and origins of constitutive IFN. Recently, lipopolysaccharide (LPS)-induced IFN was implicated as a driver of necroptosis, a necrotic form of cell death downstream of receptor-interacting protein (RIP) kinase activation and executed by mixed lineage kinase like-domain (MLKL) protein. We found that the pre-established IFN status of the cell, instead of LPS-induced IFN, is critical for the early initiation of necroptosis in macrophages. This pre-established IFN signature stems from cytosolic DNA sensing via cGAS/STING, and maintains the expression of MLKL and one or more unknown effectors above a critical threshold to allow for MLKL oligomerization and cell death. Finally, we found that elevated IFN-signaling in systemic lupus erythematosus (SLE) augments necroptosis, providing a link between pathological IFN and tissue damage during autoimmunity.