Merck
  • Dye-coupling between neonatal spinal motoneurons and interneurons revealed by prolonged back-filling of a ventral root with a low molecular weight tracer in the mouse.
Dye-coupling between neonatal spinal motoneurons and interneurons revealed by prolonged back-filling of a ventral root with a low molecular weight tracer in the mouse.
Scientific reports (2019-03-03)
Dvir Blivis, Melanie Falgairolle, Michael J O'Donovan
ABSTRACT
We investigated dye-coupling between motoneurons in the L6 segment of the neonatal mouse spinal cord that contains limb-innervating motoneurons and sexually dimorphic motor nuclei. Using an isolated spinal cord preparation, we back-filled the cut, L6 ventral root with the small molecule Neurobiotin, or the much larger dextran-conjugated fluorophores for 16-24 hours. Motoneurons and parasympathetic preganglionic neurons were filled with both markers, but dye-coupling was only seen with Neurobiotin fills. Following a neurobiotin fill, fluorescence was observed in contralateral motoneurons, in motoneurons innervating adjacent ventral roots, and in ChAT-negative, putative interneurons outside of the motoneuron pools in addition to the directly back-labeled L6 motoneurons. It is known that the gap junction protein connexin-36 is widely expressed in the sexually dimorphic motoneurons of the L6 segment, suggesting that the dye-coupling is mediated by gap junctions. The technique has revealed previously unknown connections of motoneurons in the neonatal mouse cord that are likely to play important roles in the development and function of spinal circuits.
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat