Merck
  • Control over single-cell distribution of G1 lengths by WNT governs pluripotency.
Control over single-cell distribution of G1 lengths by WNT governs pluripotency.
PLoS biology (2019-09-27)
Jiwon Jang, Dasol Han, Mahdi Golkaram, Morgane Audouard, Guojing Liu, Daniel Bridges, Stefan Hellander, Alex Chialastri, Siddharth S Dey, Linda R Petzold, Kenneth S Kosik
ABSTRACT
The link between single-cell variation and population-level fate choices lacks a mechanistic explanation despite extensive observations of gene expression and epigenetic variation among individual cells. Here, we found that single human embryonic stem cells (hESCs) have different and biased differentiation potentials toward either neuroectoderm or mesendoderm depending on their G1 lengths before the onset of differentiation. Single-cell variation in G1 length operates in a dynamic equilibrium that establishes a G1 length probability distribution for a population of hESCs and predicts differentiation outcome toward neuroectoderm or mesendoderm lineages. Although sister stem cells generally share G1 lengths, a variable proportion of cells have asymmetric G1 lengths, which maintains the population dispersion. Environmental Wingless-INT (WNT) levels can control the G1 length distribution, apparently as a means of priming the fate of hESC populations once they undergo differentiation. As a downstream mechanism, global 5-hydroxymethylcytosine levels are regulated by G1 length and thereby link G1 length to differentiation outcomes of hESCs. Overall, our findings suggest that intrapopulation heterogeneity in G1 length underlies the pluripotent differentiation potential of stem cell populations.
Sigma-Aldrich
Anti-Beta Catenin Antibody, clone 7F7.2, clone 7F7.2, Upstate®, from mouse
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid