Merck
  • Global deletion of Optineurin results in altered type I IFN signaling and abnormal bone remodeling in a model of Paget's disease.
Global deletion of Optineurin results in altered type I IFN signaling and abnormal bone remodeling in a model of Paget's disease.
Cell death and differentiation (2019-05-12)
Sing-Wai Wong, Bo-Wen Huang, Xiangxiang Hu, Eui Ho Kim, Joseph P Kolb, Ricardo J Padilla, Peng Xue, Lufei Wang, Thomas H Oguin, Patricia A Miguez, Henry C Tseng, Ching-Chang Ko, Jennifer Martinez
ABSTRACT
Genome-wide association studies (GWAS) have identified Optineurin (OPTN) as genetically linked to Paget's disease of the bone (PDB), a chronic debilitating bone remodeling disorder characterized by localized areas of increased bone resorption and abnormal bone remodeling. However, only ~10% of mouse models with a mutation in Optn develop PDB, thus hindering the mechanistic understanding of the OPTN-PDB axis. Here, we reveal that 100% of aged Optn global knockout (Optn-/-) mice recapitulate the key clinical features observed in PDB patients, including polyostotic osteolytic lesions, mixed-phase lesions, and increased serum levels of alkaline phosphatase (ALP). Differentiation of primary osteoclasts ex vivo revealed that the absence of Optn resulted in an increased osteoclastogenesis. Mechanistically, Optn-deficient osteoclasts displayed a significantly decreased type I interferon (IFN) signature, resulting from both defective production of IFNβ and impaired signaling via the IFNα/βR, which acts as a negative feedback loop for osteoclastogenesis and survival. These data highlight the dual roles of OPTN in the type I IFN response to restrain osteoclast activation and bone resorption, offering a novel therapeutic target for PDB. Therefore, our study describes a novel and essential mouse model for PDB and define a key role for OPTN in osteoclast differentiation.
Sigma-Aldrich
Anti-Huntingtin Protein Antibody, a.a. 181-810, clone 1HU-4C8, ascites fluid, clone 1HU-4C8, Chemicon®
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)