Merck
  • MAGE-A1 in lung adenocarcinoma as a promising target of chimeric antigen receptor T cells.
MAGE-A1 in lung adenocarcinoma as a promising target of chimeric antigen receptor T cells.
Journal of hematology & oncology (2019-10-24)
Yuan Mao, Weifei Fan, Hao Hu, Louqian Zhang, Jerod Michel, Yaqin Wu, Jun Wang, Lizhou Jia, Xiaojun Tang, Li Xu, Yan Chen, Jin Zhu, Zhenqing Feng, Lin Xu, Rong Yin, Qi Tang
ABSTRACT
Cancer/testis antigens (CTAs) are a special type of tumor antigen and are believed to act as potential targets for cancer immunotherapy. In this study, we first screened a rational CTA MAGE-A1 for lung adenocarcinoma (LUAD) and explored the detailed characteristics of MAGE-A1 in LUAD development through a series of phenotypic experiments. Then, we developed a novel MAGE-A1-CAR-T cell (mCART) using lentiviral vector based on our previous MAGE-A1-scFv. The anti-tumor effects of this mCART were finally investigated in vitro and in vivo. The results showed striking malignant behaviors of MAGE-A1 in LUAD development, which further validated the rationality of MAGE-A1 as an appropriate target for LUAD treatment. Then, the innovative mCART was successfully constructed, and mCART displayed encouraging tumor-inhibitory efficacy in LUAD cells and xenografts. Taken together, our data suggest that MAGE-A1 is a promising candidate marker for LUAD therapy and the MAGE-A1-specific CAR-T cell immunotherapy may be an effective strategy for the treatment of MAGE-A1-positive LUAD.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%