Merck
  • Furin inhibition prevents hypoxic and TGFβ-mediated blood-brain barrier disruption.
Furin inhibition prevents hypoxic and TGFβ-mediated blood-brain barrier disruption.
Experimental cell research (2019-07-25)
Julia Baumann, Sheng-Fu Huang, Max Gassmann, Chih-Chieh Tsao, Omolara O Ogunshola
ABSTRACT
Hypoxic blood-brain barrier (BBB) dysfunction is a common feature of CNS diseases however mechanisms underlying barrier disturbance are still largely unknown. This study investigated the role of transforming growth factor β (TGFβ), a cytokine known to induce expression of the proprotein convertase Furin, in hypoxia-mediated barrier compromise. We show that exposure of brain endothelial cells (ECs) to hypoxia (1% O2) rapidly stimulates their migration. Additional exogenous TGFβ (0.4 nM) exposure potentiated this effect and increased Furin expression in a TGFβ type I receptor activin-like kinase 5 (ALK5) - dependent manner (prevented by 10 μM SB431542). Furin inhibition prevented hypoxia-induced EC migration and blocked TGFβ-induced potentiation suggesting existence of a feedback loop. TGFβ and Furin were also critical for hypoxia-induced BBB dysfunction. TGFβ treatment aggravated hypoxia-induced BBB permeability but ALK5 or Furin blockade reversed injury-induced permeability changes. Thus during insult Furin compromises endothelial integrity by mediating the effects of TGFβ. Targeting the Furin or ALK5 pathway may offer novel therapeutic strategies for improving BBB stability and CNS function during disease.
Sigma-Aldrich
Naphthofluorescein, suitable for fluorescence
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid