Merck
  • PC-1 NF suppresses high glucose-stimulated inflammation and extracellular matrix accumulation in glomerular mesangial cells via the Wnt/β-catenin signaling.
PC-1 NF suppresses high glucose-stimulated inflammation and extracellular matrix accumulation in glomerular mesangial cells via the Wnt/β-catenin signaling.
Experimental and therapeutic medicine (2019-08-15)
Liangxiang Xiao, Yingying Chen, Yang Yuan, Bo Xu, Qing Gao, Ping Chen, Tianying Zhang, Tianjun Guan
ABSTRACT
Diabetic nephropathy (DN) is the leading cause of end-stage renal disease worldwide with high morbidity and mortality. Glomerular mesangial cell (MC) proliferation, inflammatory cell infiltration and extracellular matrix (ECM) accumulation are the main pathological characteristics of DN. A previous study revealed that polycystin-1 N-terminal fragment (PC-1 NF) fusion protein could inhibit ECM accumulation in a mesangial proliferative glomerulonephritis model. However, the role of PC-1 NF fusion protein in DN remains unknown. The results of the present study indicated that PC-1 NF fusion protein significantly abolished high glucose (HG)-induced glomerular MC viability over three time points measured (24, 48 and 72 h). In addition, PC-1 NF suppressed the levels of monocyte chemotactic peptide-1 and tumor necrosis factor α, as well as the expression of fibronectin and collagen IV, in HG-stimulated MCs. Furthermore, PC-1 NF fusion protein efficiently inhibited the activation of Wnt/β-catenin signaling pathway in HG-stimulated MCs. Taken together, these data indicated that PC-1 NF fusion protein inhibited HG-induced MC proliferation, inflammation, and ECM expression via the modulation of the Wnt signaling pathway. The present study indicated that PC-1 NF fusion protein may be a potential agent in treating DN.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution