Merck
  • Prediction of ALK mutations mediating ALK-TKIs resistance and drug re-purposing to overcome the resistance.
Prediction of ALK mutations mediating ALK-TKIs resistance and drug re-purposing to overcome the resistance.
EBioMedicine (2019-01-22)
Koutaroh Okada, Mitsugu Araki, Takuya Sakashita, Biao Ma, Ryo Kanada, Noriko Yanagitani, Atsushi Horiike, Sumie Koike, Tomoko Oh-Hara, Kana Watanabe, Keiichi Tamai, Makoto Maemondo, Makoto Nishio, Takeshi Ishikawa, Yasushi Okuno, Naoya Fujita, Ryohei Katayama
ABSTRACT
Alectinib has shown a greater efficacy to ALK-rearranged non-small-cell lung cancers in first-line setting; however, most patients relapse due to acquired resistance, such as secondary mutations in ALK including I1171N and G1202R. Although ceritinib or lorlatinib was shown to be effective to these resistant mutants, further resistance often emerges due to ALK-compound mutations in relapse patients following the use of ceritinib or lorlatinib. However, the drug for overcoming resistance has not been established yet. We established lorlatinib-resistant cells harboring ALK-I1171N or -G1202R compound mutations by performing ENU mutagenesis screening or using an in vivo mouse model. We performed drug screening to overcome the lorlatinib-resistant ALK-compound mutations. To evaluate these resistances in silico, we developed a modified computational molecular dynamic simulation (MP-CAFEE). We identified 14 lorlatinib-resistant ALK-compound mutants, including several mutants that were recently discovered in lorlatinib-resistant patients. Some of these compound mutants were found to be sensitive to early generation ALK-TKIs and several BCR-ABL inhibitors. Using our original computational simulation, we succeeded in demonstrating a clear linear correlation between binding free energy and in vitro experimental IC50 value of several ALK-TKIs to single- or compound-mutated EML4-ALK expressing Ba/F3 cells and in recapitulating the tendency of the binding affinity reduction by double mutations found in this study. Computational simulation revealed that ALK-L1256F single mutant conferred resistance to lorlatinib but increased the sensitivity to alectinib. We discovered lorlatinib-resistant multiple ALK-compound mutations and an L1256F single mutation as well as the potential therapeutic strategies for these ALK mutations. Our original computational simulation to calculate the binding affinity may be applicable for predicting resistant mutations and for overcoming drug resistance in silico. FUND: This work was mainly supported by MEXT/JSPS KAKENHI Grants and AMED Grants.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse