Merck
  • De novo phosphorylation of H2AX by WSTF regulates transcription-coupled homologous recombination repair.
De novo phosphorylation of H2AX by WSTF regulates transcription-coupled homologous recombination repair.
Nucleic acids research (2019-05-03)
Jae-Hoon Ji, Sunwoo Min, Sunyoung Chae, Geun-Hyoung Ha, Yonghyeon Kim, Yeon-Ji Park, Chang-Woo Lee, Hyeseong Cho
ABSTRACT
Histone H2AX undergoes a phosphorylation switch from pTyr142 (H2AX-pY142) to pSer139 (γH2AX) in the DNA damage response (DDR); however, the functional role of H2AX-pY142 remains elusive. Here, we report a new layer of regulation involving transcription-coupled H2AX-pY142 in the DDR. We found that constitutive H2AX-pY142 generated by Williams-Beuren syndrome transcription factor (WSTF) interacts with RNA polymerase II (RNAPII) and is associated with RNAPII-mediated active transcription in proliferating cells. Also, removal of pre-existing H2AX-pY142 by ATM-dependent EYA1/3 phosphatases disrupts this association and requires for transcriptional silencing at transcribed active damage sites. The following recovery of H2AX-pY142 via translocation of WSTF to DNA lesions facilitates transcription-coupled homologous recombination (TC-HR) in the G1 phase, whereby RAD51 loading, but not RPA32, utilizes RNAPII-dependent active RNA transcripts as donor templates. We propose that the WSTF-H2AX-RNAPII axis regulates transcription and TC-HR repair to maintain genome integrity.
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Sigma-Aldrich
α-Amanitin, from Amanita phalloides, ≥85% (HPLC), powder