Merck
  • Mixture of Azolium Tetraphenylborate with Isopropylthioxanthone: A New Class of N-Heterocyclic Carbene (NHC) Photogenerator for Polyurethane, Polyester, and ROMP Polymers Synthesis.
Mixture of Azolium Tetraphenylborate with Isopropylthioxanthone: A New Class of N-Heterocyclic Carbene (NHC) Photogenerator for Polyurethane, Polyester, and ROMP Polymers Synthesis.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-04-26)
Thi Kim Hoang Trinh, Jean-Pierre Malval, Fabrice Morlet-Savary, Julien Pinaud, Patrick Lacroix-Desmazes, Corine Reibel, Valérie Héroguez, Abraham Chemtob
ABSTRACT
In the search of smarter routes to control the conditions of N-heterocyclic carbene (NHCs) formation, a two-component air-stable NHC photogenerating system is reported. It relies on the irradiation at 365 nm of a mixture of 2-isopropylthioxanthone (ITX) with 1,3-bis(mesityl)imidazoli(ni)um tetraphenylborate. The photoinduced liberation of NHC is evidenced by reaction with a mesitoyl radical to form an NHC-radical adduct detectable by electron spin resonance spectroscopy. The NHC yield can be determined by 1 H NMR spectroscopy through the formation of a soluble and stable NHC-carbodiimide adduct. To deprotonate the azolium salt and liberate the NHC, a mechanism is proposed in which the role of base is played by ITX radical anion formed in situ by a primary photoinduced electron-transfer reaction between electronically excited ITX (oxidant) and BPh4- (reductant). An NHC yield as high as 70 % is achieved upon starting with a stoichiometric ratio of ITX and azolium salt. Three different photoNHC-mediated polymerizations are described: synthesis of polyurethane and polyester by organocatalyzed step-growth polymerization and ring-opening copolymerization, respectively, and generation of polynorbornene by ring-opening metathesis polymerization using an NHC-coordinated Ru catalyst formed in situ.