Merck
  • Dimerization of MICU Proteins Controls Ca2+ Influx through the Mitochondrial Ca2+ Uniporter.
Dimerization of MICU Proteins Controls Ca2+ Influx through the Mitochondrial Ca2+ Uniporter.
Cell reports (2019-01-31)
Yangfei Xing, Mingfei Wang, Jia Wang, Zhenzhen Nie, Guangyan Wu, Xue Yang, Yuequan Shen
ABSTRACT
The mitochondrial Ca2+ uniporter complex (MCUC) is responsible for Ca2+ influx into the mitochondrial matrix, playing critical roles in various mitochondrial functions. Eukaryotic MCUC consists of multiple subunits, and its Ca2+ influx activity is controlled by regulatory subunits, including mitochondrial Ca2+ uptake 1 (MICU1) and its paralogs (MICU2 and MICU3). However, the underlying mechanism remains unclear. Here, we determined multiple crystal structures of MICU2 and MICU3 from Homo sapiens. Our data demonstrate that distinct MICU protein N-domains determine the specific type of MICU dimers that perform the opposing roles in mitochondrial Ca2+ uptake at low cytosolic Ca2+ levels. In contrast, at high cytosolic Ca2+ levels, all MICU proteins undergo dimer rearrangement induced by Ca2+ binding, which releases the suppression of the MCUC pore-forming subunit and promotes the influx of large amounts of Ca2+. Altogether, our results elucidate the delicate mechanism of mitochondrial Ca2+ uptake regulation by MICU proteins.
Sigma-Aldrich
Anti-MICU2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-c-Myc antibody produced in mouse, clone 9E10, ascites fluid