- Dietary Branched-Chain Amino Acids Regulate Food Intake Partly through Intestinal and Hypothalamic Amino Acid Receptors in Piglets.
Dietary Branched-Chain Amino Acids Regulate Food Intake Partly through Intestinal and Hypothalamic Amino Acid Receptors in Piglets.
Strategies to increase feed intake are of great importance for producing more meat in swine production. Intestinal and hypothalamic amino acid receptors are found to largely participate in feed intake regulation. The purpose of the current research is to study the function of branched-chain amino acid (BCAA) supplementation in the regulation of feed intake through sensors that can detect amino acids in piglets. Twenty-four piglets were assigned one of four treatments and fed one of the experimental diets for either a short period (Expt. 1) or a long period (Expt. 2): a normal protein diet (NP, 20.04% CP), a reduced-protein diet (RP, 17.05% CP), or a reduced-protein test diet supplemented with one of two doses of BCAAs (BCAA1, supplemented with 0.13% l-isoleucine, 0.09% l-leucine, and 0.23% l-valine; BCAA2, supplemented with the 150% standardized ileal digestibility BCAA requirement, as recommended by the National Research Council (2012)). In Expt. 1, no differences were observed in the feed intake among piglets fed different diets ( P > 0.05). In Expt. 2, compared with the RP group, the feed intake of piglets was significantly increased after sufficient BCAAs were supplemented in the BCAA1 group, which was associated with decreased cholecystokinin secretion ( P < 0.05), down-regulated expression of type-1 taste receptors 1/3 (T1R1/T1R3) in the intestine, as well as increased expression of pro-opiomelanocortin, activated general control nonderepressible 2 (GCN2), and eukaryotic initiation factor 2α (eIF2α) in the hypothalamus ( P < 0.05). However, the feed intake was decreased for unknown reasons when the piglets were fed a BCAA over-supplemented diet. Our study confirmed that a BCAA-deficient diet inhibited feed intake through two potential ways: regulating the amino acid T1R1/T1R3 receptor in the intestine or activating GCN2/eIF2α pathways in the hypothalamus.