Merck
  • l-Arginine Synthesis from l-Citrulline in Myeloid Cells Drives Host Defense against Mycobacteria In Vivo.
l-Arginine Synthesis from l-Citrulline in Myeloid Cells Drives Host Defense against Mycobacteria In Vivo.
Journal of immunology (Baltimore, Md. : 1950) (2019-02-03)
Shannon M Lange, Melanie C McKell, Stephanie M Schmidt, Junfang Zhao, Rebecca R Crowther, Lisa C Green, Rebecca L Bricker, Eusondia Arnett, S Eleonore Köhler, Larry S Schlesinger, Kenneth D R Setchell, Joseph E Qualls
ABSTRACT
Immunonutrition as a therapeutic approach is rapidly gaining interest in the fight against infection. Targeting l-arginine metabolism is intriguing, considering this amino acid is the substrate for antimicrobial NO production by macrophages. The importance of l-arginine during infection is supported by the finding that inhibiting its synthesis from its precursor l-citrulline blunts host defense. During the first few weeks following pulmonary mycobacterial infection, we found a drastic increase in l-citrulline in the lung, even though serum concentrations were unaltered. This correlated with increased gene expression of the l-citrulline-generating (i.e., iNOS) and l-citrulline-using (i.e., Ass1) enzymes in key myeloid populations. Eliminating l-arginine synthesis from l-citrulline in myeloid cells via conditional deletion of either Ass1 or Asl resulted in increased Mycobacterium bovis bacillus Calmette-Guérin and Mycobacterium tuberculosis H37Rv burden in the lungs compared with controls. Our data illustrate the necessity of l-citrulline metabolism for myeloid defense against mycobacterial infection and highlight the potential for host-directed therapy against mycobacterial disease targeting this nutrient and/or its metabolic pathway.