- Deletion of a conserved transcript PG_RS02100 expressed during logarithmic growth in Porphyromonas gingivalis results in hyperpigmentation and increased tolerance to oxidative stress.
Deletion of a conserved transcript PG_RS02100 expressed during logarithmic growth in Porphyromonas gingivalis results in hyperpigmentation and increased tolerance to oxidative stress.
The oral obligate anaerobe Porphyromonas gingivalis possesses a small conserved transcript PG_RS02100 of unknown function we previously identified using small RNA-seq analysis as expressed during logarithmic growth. In this study, we sought to determine if PG_RS02100 plays a role in P. gingivalis growth or stress response. We show that a PG_RS02100 deletion mutant's (W83Δ514) ability to grow under anaerobic conditions was no different than wildtype (W83), but it was better able to survive hydrogen peroxide exposure when cultured under heme limiting growth conditions, and was more aerotolerant when plated on enriched whole blood agar and exposed to atmospheric oxygen. Together, these results indicate that PG_RS02100 plays a role in surviving oxidative stress in actively growing P. gingivalis and that P. gingivalis' response to exogenous hydrogen peroxide stress is linked to heme availability. Relative qRT-PCR expression analysis of oxyR, trx-1, tpx, sodB, ahpC, dinF, cydB, and frd, in W83Δ514 and W83 in response to 1 h exogenous dioxygen or hydrogen peroxide exposure, when cultured with varying heme availability, support our phenotypic evidence that W83Δ514 has a more highly primed defense system against exogenous peroxide, dioxygen, and heme generated ROS. Interestingly, W83Δ514 turned black faster than W83 when cultured on whole blood agar, suggesting it was able to accumulate heme more rapidly. The mechanism of increased heme acquisition observed in W83Δ514 is not yet known. However, it is clear that PG_RS02100 is involved in modulating the P. gingivalis cell surface in a manner related to survival, particularly against oxidative stress.