Merck
Retrieval of weak x-ray scattering using edge illumination.
Optics letters (2018-08-15)
Charlotte J Maughan Jones, Fabio A Vittoria, Alessandro Olivo, Marco Endrizzi, Peter R T Munro
ABSTRACT
X-ray phase contrast imaging provides additional modes of image contrast compared to conventional attenuation-based x-ray imaging, thus providing additional structural and functional information about the sample. The edge-illumination (EI) technique has been used to provide attenuation, refraction, and scattering contrast in both biological and non-biological samples. However, the retrieval of low scattering signals by fitting a single Gaussian remains problematic, principally due to the inability of the EI system to achieve perfect dark-field illumination. We present a new retrieval method that fits three Gaussians, which successfully overcomes this limitation, and provide examples of the retrieval of such signals in highly absorbing, weakly scattering samples.