Merck
  • Evaluation of the Dietary Supplementation of a Formulation Containing Ascorbic Acid and a Solid Dispersion of Curcumin with Boric Acid against Salmonella Enteritidis and Necrotic Enteritis in Broiler Chickens.
Evaluation of the Dietary Supplementation of a Formulation Containing Ascorbic Acid and a Solid Dispersion of Curcumin with Boric Acid against Salmonella Enteritidis and Necrotic Enteritis in Broiler Chickens.
Animals : an open access journal from MDPI (2019-04-25)
Daniel Hernandez-Patlan, Bruno Solís-Cruz, Karine Patrin Pontin, Juan D Latorre, Mikayla F A Baxter, Xochitl Hernandez-Velasco, Ruben Merino-Guzman, Abraham Méndez-Albores, Billy M Hargis, Raquel Lopez-Arellano, Guillermo Tellez-Isaias
ABSTRACT
Two experiments were conducted to evaluate the effect of the prophylactic or therapeutic administration of a 0.1% mixture containing ascorbic acid and a solid dispersion of curcumin with polyvinylpyrrolidone and boric acid (AA-CUR/PVP-BA) against Salmonella Enteritidis (S. Enteritidis) in broiler chickens. A third experiment was conducted to evaluate the impact of the dietary administration of 0.1% AA-CUR/PVP-BA in a necrotic enteritis (NE) model in broiler chickens. The prophylactic administration of 0.1% AA-CUR/PVP-BA significantly decreased S. Enteritidis colonization in cecal tonsils (CT) when compared to the positive control group (PC, p < 0.05). The therapeutic administration of 0.1% AA-CUR/PVP-BA significantly reduced the concentration of S. Enteritidis by 2.05 and 2.71 log in crop and CT, respectively, when compared with the PC on day 10 post-S. Enteritidis challenge. Furthermore, the serum FITC-d concentration and total intestinal IgA levels were also significantly lower in chickens that received 0.1% AA-CUR/PVP-BA. Contrary, the PC group showed significantly higher total intestinal IgA levels compared to the negative control or AA-CUR/PVP-BA groups in the NE model. However, 0.1% AA-CUR/PVP-BA showed a better effect in reducing the concentration of S. Enteritidis when compared to the NE model. Further studies with higher concentration of AA-CUR/PVP-BA into the feed to extend these preliminary results are currently being evaluated.