Merck
G proteins.
Trends in biochemical sciences (1992-10-01)
J R Hepler, A G Gilman
ABSTRACT
The family of heterotrimeric guanine nucleotide-binding regulatory proteins (G proteins) serves an essential role in transducing receptor-generated signals across the plasma membrane. Recent findings reveal unexpected functional roles for individual G protein subunits. Thus, GTP-binding alpha-subunits and the beta gamma-subunit complex can influence the activity of effector molecules independently or simultaneously, either synergistically or in opposition, to elicit a complex constellation of cellular events.
Sigma-Aldrich
Guanosine 5′-[β,γ-imido]triphosphate trisodium salt hydrate, ≥85% (HPLC), powder