Merck
  • Label-free emission assay of mercuric ions using DNA duplexes of poly(dT).
Label-free emission assay of mercuric ions using DNA duplexes of poly(dT).
Dalton transactions (Cambridge, England : 2003) (2011-05-13)
Byul Nim Oh, Sangji Park, Jun Ren, Yoon Jung Jang, Seog K Kim, Jinheung Kim
ABSTRACT
In this study, an assay to quantify the presence of mercuric ions and methyl mercury by double-stranded DNA containing a poly(dT) sequence was developed using a light switch compound, Ru(phen)(2)(dppz)(2+) (1), which is known to intercalate into double-stranded DNA. Upon treatment with mercuric ions, the metal-to-ligand charge transfer (MLCT) emission derived from the intercalation of 1 was reduced due to the formation of DNA duplexes containing T-Hg(2+)-T base pairs by the dehybridization of poly(dT)-poly(dA) duplexes at room temperature. As the concentration of Hg(2+) was increased, the emission of 1 gradually decreased. This label-free method had a detection limit of 5 nM. Other metal ions, such as K(+), Ag(+), Ca(2+), Mg(2+), Zn(2+), Mn(2+), Co(2+), Ni(2+), Cu(2+), Cd(2+), Cr(3+), Fe(3+), had no significant effect on reducing emission. This emission method can differentiate matched and mismatched poly(dT) sequences based on the dehybridization rate of dsDNA and the rate decreased in the order of T(10)C·A(11)∼ T(10)A·A(11) > T(10)G·A(11) > T(11)·A(11).
Sigma-Aldrich
Polythymidylic acid sodium salt, homopolymer