Merck
Active Targeting of the Nucleus Using Nonpeptidic Boronate Tags.
Journal of the American Chemical Society (2017-06-10)
Rui Tang, Ming Wang, Moumita Ray, Ying Jiang, Ziwen Jiang, Qiaobing Xu, Vincent M Rotello
ABSTRACT
Active intracellular transport is a central mechanism in cell biology, directed by a limited set of naturally occurring signaling peptides. Here, we report the first nonpeptide moiety that recruits intracellular transport machinery for nuclear targeting. Proteins synthetically modified with a simple aromatic boronate motif are actively trafficked to the nucleus via the importin α/β pathway. Significantly, proteins too large to passively diffuse through nuclear pores were readily imported into the nucleus through this boronate-mediated pathway. The use of this simple motif to provide active intracellular targeting provides a promising strategy for directing subcellular localization for therapeutic and fundamental applications.
Sigma-Aldrich
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl (bromoethyl)carbamate
Sigma-Aldrich
2-((4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)oxy)ethan-1-amine, ≥95%