Merck
  • Respiratory syncytial virus G and/or SH protein alters Th1 cytokines, natural killer cells, and neutrophils responding to pulmonary infection in BALB/c mice.
Respiratory syncytial virus G and/or SH protein alters Th1 cytokines, natural killer cells, and neutrophils responding to pulmonary infection in BALB/c mice.
Journal of virology (1999-08-10)
R A Tripp, D Moore, L Jones, W Sullender, J Winter, L J Anderson
ABSTRACT
BALB/c mice sensitized to vaccinia virus expressed G protein of respiratory syncytial virus (RSV) develop a Th2-type cytokine response and pulmonary eosinophilia when challenged with live RSV. In this study, BALB/c mice were immunized or challenged with an RSV mutant lacking the G and SH proteins or with DNA vaccines coding for RSV G or F protein. F or G protein DNA vaccines were capable of sensitizing for pulmonary eosinophilia. The absence of the G and/or SH protein in the infecting virus resulted in a consistent increase both in pulmonary natural killer cells and in gamma interferon and tumor necrosis factor expression, as well as, with primary infection, a variable increase in neutrophils and CD11b(+) cells. The development of pulmonary eosinophilia in formalin-inactivated RSV-vaccinated mice required the presence of the G and/or SH protein in the challenge virus. These data show that G and/or SH protein has a marked impact on the inflammatory and innate immune response to RSV infection.
Sigma-Aldrich
Anti-RSV Antibody, fusion protein, all type A, B strains, clone 131-2A, clone 131-2A, Chemicon®, from mouse